Assay Details
Target Gene Details
Entrez Gene ID: | 267 |
Gene Name: | autocrine motility factor receptor |
Gene Aliases: |
GP78, RNF45 |
Location: |
Chr.16:56361452-56425538 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AMFR | NM_001144.5 | 9 | 1338 | NP_001135.3 |
| NM_001323511.1 | 9 | 957 | NP_001310440.1 | |
| NM_001323512.1 | 9 | 1338 | NP_001310441.1 | |
| XM_005255890.3 | 9 | 1398 | XP_005255947.1 | |
| AF124145.1 | 9 | 1192 | AAD56722.1 | |
| AK023874.1 | ||||
| BC017043.1 | 2 | 75 | AAH17043.1 | |
| BC069197.1 | 9 | 1338 | AAH69197.1 | |
| BI766313.1 | 3 | 272 | ||
| CD642692.1 | 3 | 228 | ||
| L35233.1 | 2 | 71 | AAA79362.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833242 | Chr.16:56259354 - 56446840 on Build GRCh38 | Loss |
|
| nsv833243 | Chr.16:56375371 - 56549461 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map