Assay Details
Target Gene Details
Entrez Gene ID: | 100528018 |
Gene Name: | ARL2-SNX15 readthrough (NMD candidate) |
Gene Aliases: |
- |
Location: |
Chr.11:65014113-65040572 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ARL2-SNX15 | NR_037650.1 | 6 | 842 | |
| AK096447.1 | 6 | 776 |
Target Gene Details
Entrez Gene ID: | 29907 |
Gene Name: | sorting nexin 15 |
Gene Aliases: |
HSAF001435 |
Location: |
Chr.11:65027408-65040572 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SNX15 | NM_013306.4 | 3 | 318 | NP_037438.2 |
| NM_147777.3 | 3 | 318 | NP_680086.2 | |
| AB209439.1 | 3 | 186 | BAD92676.1 | |
| AF001435.1 | 3 | 244 | AAB81205.1 | |
| AF175267.2 | 3 | 227 | AAF89955.1 | |
| AF175268.1 | 3 | 188 | AAF89956.1 | |
| AK315034.1 | 3 | 249 | ||
| AK315642.1 | 3 | 259 | ||
| BC009897.2 | 3 | 236 | AAH09897.1 | |
| BC012767.2 | 3 | 288 | AAH12767.1 | |
| BC014520.2 | 3 | 288 | AAH14520.1 | |
| BT006631.1 | 3 | 188 | AAP35277.1 | |
| DA079695.1 | 3 | 319 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3626669 | Chr.11:65023467 - 65037106 on Build GRCh38 | Gain |
|
| nsv526181 | Chr.11:64870569 - 65489858 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map