Assay Details
Target Gene Details
Entrez Gene ID: | 23408 |
Gene Name: | sirtuin 5 |
Gene Aliases: |
SIR2L5 |
Location: |
Chr.6:13574222-13615158 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SIRT5 | NM_001193267.2 | 5 | 701 | NP_001180196.1 |
| NM_001242827.1 | 4 | 567 | NP_001229756.1 | |
| NM_012241.4 | 5 | 723 | NP_036373.1 | |
| NM_031244.3 | 5 | 701 | NP_112534.1 | |
| XM_005248968.4 | 5 | 638 | XP_005249025.1 | |
| XM_011514444.2 | 6 | 772 | XP_011512746.1 | |
| XM_017010620.1 | 4 | 601 | XP_016866109.1 | |
| XM_017010621.1 | 5 | 638 | XP_016866110.1 | |
| XM_017010622.1 | 6 | 831 | XP_016866111.1 | |
| XM_017010623.1 | 6 | 961 | XP_016866112.1 | |
| XM_017010624.1 | 4 | 601 | XP_016866113.1 | |
| XM_017010625.1 | 4 | 601 | XP_016866114.1 | |
| AF083110.1 | 5 | 684 | AAD40853.1 | |
| AJ420496.1 | 5 | 630 | ||
| AK000355.1 | 5 | 629 | BAA91107.1 | |
| AK294162.1 | 4 | 495 | ||
| AK302467.1 | 5 | 636 | ||
| BC000126.1 | 5 | 707 | AAH00126.1 | |
| BX538030.1 | 4 | 514 | CAD97975.1 | |
| CB155518.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3608129 | Chr.6:13525301 - 13607816 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map