Assay Details
Target Gene Details
Entrez Gene ID: | 100499484 |
Gene Name: | SUGT1-1300002K09Rik pseudogene |
Gene Aliases: |
- |
Location: |
Chr.9:97238426-97297314 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC100499484 | NR_036526.1 | 5 | 378 | |
| AK301827.1 | 5 | 321 | ||
| AK303497.1 | 5 | 355 | ||
| DA153387.1 | 5 | 379 |
Target Gene Details
Entrez Gene ID: | 57653 |
Gene Name: | LOC100499484-C9orf174 readthrough |
Gene Aliases: |
- |
Location: |
Chr.9:97238426-97377295 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC100499484-C9ORF174 | NR_036527.1 | 5 | 378 | |
| NR_036528.1 | 5 | 378 | ||
| NR_036529.1 | 5 | 378 |

Genomic Map