Assay Details
Target Gene Details
Entrez Gene ID: | 79841 |
Gene Name: | ATP/GTP binding protein like 2 |
Gene Aliases: |
CCP2 |
Location: |
Chr.11:47659168-47715397 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 18 - Exon 18 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv469858 | Chr.11:47625963 - 47815620 on Build GRCh38 | Gain |
|
| dgv1111n100 | Chr.11:47653918 - 48644472 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map