Assay Details
Target Gene Details
Entrez Gene ID: | 114780 |
Gene Name: | polycystin 1 like 2 (gene/pseudogene) |
Gene Aliases: |
PC1L2 |
Location: |
Chr.16:81100879-81220370 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 39 - Exon 39 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PKD1L2 | NM_001278425.1 | NP_001265354.1 | ||
| NM_052892.3 | NP_443124.3 | |||
| NR_126532.1 | ||||
| AK131378.1 | BAD18529.1 | |||
| AY164483.1 | AAO32796.1 | |||
| AY371495.1 | AAQ73173.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv520035 | Chr.16:81112070 - 81123916 on Build GRCh38 | Loss |
|
| esv2751615 | Chr.16:81059494 - 81273994 on Build GRCh38 | Gain |
|
| nsv1056843 | Chr.16:81120843 - 81139057 on Build GRCh38 | Loss |
|
| nsv1067144 | Chr.16:81095693 - 81148036 on Build GRCh38 | Loss |
|
| dgv510e199 | Chr.16:81120968 - 81123769 on Build GRCh38 | Deletion |
|
| esv3639311 | Chr.16:81120941 - 81123631 on Build GRCh38 | Loss |
|
| nsv1063460 | Chr.16:81105468 - 81123780 on Build GRCh38 | Gain |
|
| esv2422360 | Chr.16:81032428 - 81147122 on Build GRCh38 | Duplication |
|
More Information
Set Membership: |
|

Genomic Map