Assay Details
Target Gene Details
Entrez Gene ID: | 2824 |
Gene Name: | glycoprotein M6B |
Gene Aliases: |
M6B |
Location: |
Chr.X:13770922-13938824 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GPM6B | NM_001001994.2 | NP_001001994.1 | ||
| NM_001001995.2 | NP_001001995.1 | |||
| NM_001001996.2 | NP_001001996.1 | |||
| NM_001318729.1 | NP_001305658.1 | |||
| NM_005278.4 | NP_005269.1 | |||
| XM_005274489.4 | XP_005274546.1 | |||
| XM_011545497.2 | XP_011543799.1 | |||
| XM_017029432.1 | XP_016884921.1 | |||
| XM_017029433.1 | XP_016884922.1 | |||
| XM_017029434.1 | XP_016884923.1 | |||
| AB209525.1 | BAD92762.1 | |||
| AK095657.1 | BAC04600.1 | |||
| AK294314.1 | ||||
| AK299675.1 | ||||
| BC008151.1 | AAH08151.1 | |||
| BC047295.1 | AAH47295.1 | |||
| BF525674.1 | ||||
| CD516484.1 | ||||
| U45955.1 | AAB16888.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv528634 | Chr.X:13696631 - 14038859 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map