Assay Details
Target Gene Details
Entrez Gene ID: | 10534 |
Gene Name: | Sjogren syndrome/scleroderma autoantigen 1 |
Gene Aliases: |
p27 |
Location: |
Chr.11:65570459-65571888 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SSSCA1 | NM_001303024.1 | NP_001289953.1 | ||
| NM_006396.2 | NP_006387.1 | |||
| AB001740.1 | BAA25263.1 | |||
| BC014791.2 | AAH14791.1 | |||
| BP351453.1 | ||||
| DR759640.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv1969n54 | Chr.11:65568234 - 65663889 on Build GRCh38 | Loss |
|
| nsv951018 | Chr.11:65525430 - 65662029 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map