Assay Details
Target Gene Details
Entrez Gene ID: | 10401 |
Gene Name: | protein inhibitor of activated STAT 3 |
Gene Aliases: |
ZMIZ5 |
Location: |
Chr.1:145848522-145859081 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PIAS3 | NM_006099.3 | 14 | 1979 | NP_006090.2 |
| AB021868.1 | 14 | 1886 | BAA78533.1 | |
| AK027557.1 | 14 | 1978 | ||
| AK125158.1 | 7 | 1756 | ||
| AL122061.1 | 7 | 963 | CAB59241.1 | |
| AL133016.1 | 4 | 1432 | CAB61357.2 | |
| BC001154.1 | 14 | 1924 | ||
| BC030556.1 | 14 | 1935 | ||
| BT007036.1 | AAP35684.1 | |||
| CR457090.1 | CAG33371.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1005055 | Chr.1:145642988 - 148991068 on Build GRCh38 | Loss |
|
| esv3587420 | Chr.1:145824369 - 145905171 on Build GRCh38 | Gain |
|
| nsv831381 | Chr.1:145784568 - 145960930 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map