Assay Details
Target Gene Details
Entrez Gene ID: | 59341 |
Gene Name: | transient receptor potential cation channel subfamily V member 4 |
Gene Aliases: |
BCYM3, CMT2C, HMSN2C, OTRPC4, SMAL, SPSMA, SSQTL1, TRP12, VRL2, VROAC |
Location: |
Chr.12:109783087-109833407 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 14 - Exon 15 |
More Information
Set Membership: |
|

Genomic Map