Assay Details
Target Gene Details
Entrez Gene ID: | 57182 |
Gene Name: | ankyrin repeat domain 50 |
Gene Aliases: |
- |
Location: |
Chr.4:124664048-124712732 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ANKRD50 | NM_001167882.1 | 4 | 4211 | NP_001161354.1 |
| NM_020337.2 | 5 | 5486 | NP_065070.1 | |
| XM_017008471.1 | 4 | 6219 | XP_016863960.1 | |
| AB033049.2 | 2 | 3430 | BAA86537.2 | |
| AK291068.1 | 5 | 5486 | ||
| AK295135.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs115876926] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map