Assay Details
Target Gene Details
Entrez Gene ID: | 22891 |
Gene Name: | zinc finger protein 365 |
Gene Aliases: |
Su48, UAN, ZNF365D |
Location: |
Chr.10:62373609-62672011 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZNF365 | NM_014951.2 | 5 | 1670 | NP_055766.2 |
| NM_199450.2 | NP_955522.1 | |||
| NM_199451.2 | NP_955523.1 | |||
| XM_017015937.1 | XP_016871426.1 | |||
| AB020651.1 | 5 | 1632 | ||
| AJ505147.1 | 5 | 1443 | CAD43726.1 | |
| AJ505148.1 | CAD43727.1 | |||
| AJ505149.1 | CAD43728.1 | |||
| AK289515.1 | 5 | 1411 | ||
| AK289808.1 | 5 | 1435 | ||
| BC035049.1 | 4 | 785 | AAH35049.1 | |
| BC060817.1 | 4 | 780 | AAH60817.1 | |
| BX647853.1 | 5 | 1463 | ||
| BX647904.1 | 5 | 1463 | CAI46100.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv8681 | Chr.10:61896798 - 62671529 on Build GRCh38 | Gain |
|
| nsv825408 | Chr.10:62399511 - 62400673 on Build GRCh38 | Loss |
|
| nsv467293 | Chr.10:62396351 - 62464341 on Build GRCh38 | Gain |
|
| nsv1035322 | Chr.10:62377308 - 62497169 on Build GRCh38 | Gain |
|
| esv34099 | Chr.10:62021186 - 62427805 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map