Assay Details
Target Gene Details
Entrez Gene ID: | 79596 |
Gene Name: | ring finger protein 219 |
Gene Aliases: |
C13orf7 |
Location: |
Chr.13:78614286-78659179 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RNF219 | NM_024546.3 | 6 | 1667 | NP_078822.3 |
| XM_011535225.1 | 5 | 1517 | XP_011533527.1 | |
| XM_017020743.1 | 6 | 3462 | XP_016876232.1 | |
| AK023511.1 | 1 | 856 | BAB14594.1 | |
| AK098640.1 | 7 | 1755 | ||
| AL136651.1 | 6 | 1639 | CAB66586.3 | |
| BC136764.1 | 6 | 1662 | ||
| BC136765.1 | 6 | 1662 | ||
| CR936752.1 | 6 | 1644 | ||
| CR936754.1 | 6 | 1654 | ||
| CR936764.1 | 7 | 1571 |
Target Gene Details
Entrez Gene ID: | 100874222 |
Gene Name: | RNF219 antisense RNA 1 |
Gene Aliases: |
POU4F1-AS1 |
Location: |
Chr.13:78054855-78617325 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RNF219-AS1 | NR_047001.1 | 6 | 1246 | |
| AK095779.1 | 6 | 1246 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv339n67 | Chr.13:78560929 - 78651758 on Build GRCh38 | Gain |
|
| dgv234n27 | Chr.13:78566708 - 78642283 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map