Assay Details
Target Gene Details
Entrez Gene ID: | 8396 |
Gene Name: | phosphatidylinositol-5-phosphate 4-kinase type 2 beta |
Gene Aliases: |
PI5P4KB, PIP5K2B, PIP5KIIB, PIP5KIIbeta, PIP5P4KB |
Location: |
Chr.17:38765691-38799965 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PIP4K2B | NM_003559.4 | 10 | 1830 | NP_003550.1 |
| XM_011525326.2 | 11 | 1971 | XP_011523628.1 | |
| XM_011525327.2 | 12 | 1490 | XP_011523629.1 | |
| XM_011525330.2 | 9 | 1166 | XP_011523632.1 | |
| XM_017025196.1 | 11 | 1407 | XP_016880685.1 | |
| XM_017025197.1 | 11 | 1307 | XP_016880686.1 | |
| XM_017025198.1 | 9 | 1793 | XP_016880687.1 | |
| XM_017025199.1 | 10 | 1245 | XP_016880688.1 | |
| U85245.1 | 10 | 1829 | AAB48596.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833438 | Chr.17:38593441 - 38821266 on Build GRCh38 | Loss |
|
| nsv833439 | Chr.17:38743861 - 38923753 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map