Assay Details
Target Gene Details
Entrez Gene ID: | 11010 |
Gene Name: | GLI pathogenesis related 1 |
Gene Aliases: |
CRISP7, GLIPR, RTVP1 |
Location: |
Chr.12:75480733-75501936 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GLIPR1 | NM_006851.2 | 6 | 1762 | NP_006842.2 |
| XM_011537775.2 | 7 | 1812 | XP_011536077.1 | |
| XM_017018721.1 | 6 | 1642 | XP_016874210.1 |
Target Gene Details
Entrez Gene ID: | 11103 |
Gene Name: | KRR1, small subunit processome component homolog |
Gene Aliases: |
HRB2, RIP-1 |
Location: |
Chr.12:75497639-75511638 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KRR1 | NM_007043.6 | 10 | 1205 | NP_008974.5 |
| AK313687.1 | ||||
| BC016778.1 | 10 | 1179 | AAH16778.1 | |
| BC026107.2 | 10 | 1188 | AAH26107.1 | |
| BC033887.2 | 10 | 1179 | AAH33887.1 | |
| EF010919.1 | ||||
| U55766.1 | AAB00557.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv826439 | Chr.12:75498371 - 75499923 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map