Assay Details
Target Gene Details
Entrez Gene ID: | 64855 |
Gene Name: | family with sequence similarity 129 member B |
Gene Aliases: |
C9orf88, MEG-3, MINERVA, OC58, bA356B19.6 |
Location: |
Chr.9:127505338-127579007 on Build GRCh38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM129B | NM_001035534.2 | 14 | 2463 | NP_001030611.1 |
| NM_022833.3 | 14 | 2675 | NP_073744.2 | |
| XM_005252135.2 | 15 | 2522 | XP_005252192.2 | |
| XM_011518925.1 | 15 | 2694 | XP_011517227.1 | |
| AB210016.1 | 14 | 2413 | BAE06098.1 | |
| AF151783.1 | 14 | 2445 | AAK57556.1 | |
| AF192911.1 | 14 | 2666 | AAQ13825.1 | |
| AK023580.1 | 6 | 1474 | BAB14615.1 | |
| AK025804.1 | 2 | 831 | BAB15240.1 | |
| AK025951.1 | 1 | 267 | ||
| AL137555.1 | 13 | 2283 | CAB70809.1 | |
| BC001979.1 | 7 | 1562 | AAH01979.2 | |
| BC110345.1 | 5 | 1175 | AAI10346.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv466569 | Chr.9:127470144 - 127512011 on Build GRCh38 | Loss |
|
| nsv615368 | Chr.9:127504287 - 127547894 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map