Assay Details
Target Gene Details
Entrez Gene ID: | 88 |
Gene Name: | actinin alpha 2 |
Gene Aliases: |
CMD1AA, CMH23 |
Location: |
Chr.1:236686454-236764631 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ACTN2 | NM_001103.3 | NP_001094.1 | ||
| NM_001278343.1 | NP_001265272.1 | |||
| NM_001278344.1 | NP_001265273.1 | |||
| AB209521.1 | BAD92758.1 | |||
| AK297886.1 | ||||
| AK315250.1 | ||||
| BC047901.2 | AAH47901.2 | |||
| BC051770.1 | AAH51770.2 | |||
| DC378038.1 | ||||
| M86406.1 | AAA51583.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1005877 | Chr.1:235912335 - 237038508 on Build GRCh38 | Gain |
|
| nsv1013895 | Chr.1:236716519 - 236727481 on Build GRCh38 | Loss |
|
| esv2763410 | Chr.1:236334208 - 236735742 on Build GRCh38 | Gain |
|
| nsv509690 | Chr.1:236712865 - 236721707 on Build GRCh38 | Insertion |
|
| nsv4988 | Chr.1:236699164 - 236760383 on Build GRCh38 | Insertion |
|
| esv2759006 | Chr.1:236703536 - 236739064 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map