Assay Details
Target Gene Details
Entrez Gene ID: | 9757 |
Gene Name: | lysine methyltransferase 2B |
Gene Aliases: |
CXXC10, HRX2, MLL1B, MLL2, MLL4, TRX2, WBP-7, WBP7 |
Location: |
Chr.19:35717818-35738880 on Build GRCh38 |
Assay Gene Location: | Within Exon 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KMT2B | NM_014727.2 | 20 | 4636 | NP_055542.1 |
| XM_011527561.2 | 20 | 4507 | XP_011525863.2 | |
| XM_011527562.2 | 20 | 4837 | XP_011525864.1 | |
| XM_017027544.1 | 20 | 4837 | XP_016883033.1 | |
| XM_017027545.1 | 19 | 4550 | XP_016883034.1 | |
| XM_017027546.1 | 15 | 1758 | XP_016883035.1 | |
| AB002302.3 | 18 | 3736 | BAA20763.3 | |
| AF104918.1 | 18 | 2650 | AAD17932.1 | |
| AF186605.1 | 20 | 4308 | AAD56420.1 | |
| AJ007041.1 | 20 | 4636 | CAB45385.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv515536 | Chr.19:35728624 - 35750059 on Build GRCh38 | Loss |
|
| nsv579422 | Chr.19:35665686 - 35774606 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map