Assay Details
Target Gene Details
Entrez Gene ID: | 2203 |
Gene Name: | fructose-bisphosphatase 1 |
Gene Aliases: |
FBP |
Location: |
Chr.9:94603133-94640257 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FBP1 | NM_000507.3 | NP_000498.2 | ||
| NM_001127628.1 | NP_001121100.1 | |||
| XM_006717005.3 | XP_006717068.1 | |||
| AF073475.1 | AAC25774.1 | |||
| AK223395.1 | BAD97115.1 | |||
| AK292199.1 | ||||
| AK316583.1 | ||||
| AY544122.1 | AAT11153.1 | |||
| BC012927.1 | AAH12927.1 | |||
| D26054.1 | BAA05051.1 | |||
| D26055.1 | BAA05052.1 | |||
| D26056.1 | BAA05053.1 | |||
| L10320.1 | AAA35817.1 | |||
| M19922.1 | AAA35517.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3621113 | Chr.9:94207958 - 94890837 on Build GRCh38 | Gain |
|
| nsv466436 | Chr.9:94581338 - 94606867 on Build GRCh38 | Loss |
|
| esv3576685 | Chr.9:94439449 - 94658575 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map