Assay Details
Target Gene Details
Entrez Gene ID: | 56848 |
Gene Name: | sphingosine kinase 2 |
Gene Aliases: |
SK 2, SK-2, SPK 2, SPK-2 |
Location: |
Chr.19:48619291-48630407 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SPHK2 | NM_001204158.2 | 7 | 2300 | NP_001191087.1 |
| NM_001204159.2 | 7 | 2381 | NP_001191088.1 | |
| NM_001204160.2 | 6 | 2197 | NP_001191089.1 | |
| NM_001243876.1 | 6 | 2005 | NP_001230805.1 | |
| NM_020126.4 | 7 | 2477 | NP_064511.2 | |
| XM_006723292.1 | 6 | 2167 | XP_006723355.1 | |
| XM_011527133.1 | 7 | 2312 | XP_011525435.1 | |
| XM_011527134.1 | 6 | 2169 | XP_011525436.1 | |
| XM_017027008.1 | 5 | 2883 | XP_016882497.1 | |
| XM_017027009.1 | 6 | 1888 | XP_016882498.1 | |
| XM_017027010.1 | 7 | 2609 | XP_016882499.1 | |
| AF245447.1 | 5 | 2009 | AAF74124.1 | |
| AK000599.1 | BAA91280.1 | |||
| AK122735.1 | 8 | 2181 | ||
| AK300541.1 | ||||
| AL136701.1 | 7 | 2380 | CAB66636.1 | |
| BC006161.1 | 7 | 2477 | AAH06161.1 | |
| BC010671.1 | 6 | 2197 | AAH10671.1 | |
| DB367710.1 | 1 | 488 | ||
| EF107108.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv9739 | Chr.19:47901840 - 50143063 on Build GRCh38 | Gain+Loss |
|
| dgv3617n100 | Chr.19:48532114 - 48643732 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map