Assay Details
Target Gene Details
Entrez Gene ID: | 5501 |
Gene Name: | protein phosphatase 1 catalytic subunit gamma |
Gene Aliases: |
PP-1G, PP1C, PPP1G |
Location: |
Chr.12:110708369-110742978 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PPP1CC | NM_001244974.1 | 3 | 640 | NP_001231903.1 |
| NM_002710.3 | 3 | 640 | NP_002701.1 | |
| XM_011538504.2 | 3 | 561 | XP_011536806.1 | |
| XM_011538505.2 | 3 | 561 | XP_011536807.1 | |
| AK297882.1 | 3 | 358 | ||
| BC014073.2 | 3 | 533 | AAH14073.1 | |
| L07395.1 | 3 | 357 | AAA19823.1 | |
| X74008.1 | 3 | 523 | CAA52169.1 |
More Information
Set Membership: |
|

Genomic Map