Assay Details
Target Gene Details
Entrez Gene ID: | 26094 |
Gene Name: | DDB1 and CUL4 associated factor 4 |
Gene Aliases: |
WDR21, WDR21A |
Location: |
Chr.14:72926332-72962098 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 9 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1049819 | Chr.14:72911687 - 73164900 on Build GRCh38 | Gain |
|
| nsv1048734 | Chr.14:72817765 - 73069496 on Build GRCh38 | Gain |
|
| nsv1035460 | Chr.14:72854572 - 73897173 on Build GRCh38 | Gain |
|
| esv3892560 | Chr.14:72813150 - 72958973 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map