Assay Details
Target Gene Details
Entrez Gene ID: | 6559 |
Gene Name: | solute carrier family 12 member 3 |
Gene Aliases: |
NCC, NCCT, TSC |
Location: |
Chr.16:56865207-56915850 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC12A3 | NM_000339.2 | NP_000330.2 | ||
| NM_001126107.1 | NP_001119579.1 | |||
| NM_001126108.1 | NP_001119580.1 | |||
| XM_005256119.2 | XP_005256176.1 | |||
| AK223133.1 | ||||
| AK315298.1 | ||||
| U44128.1 | AAC50355.1 | |||
| X91220.1 | CAA62613.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv33108 | Chr.16:56857096 - 56883621 on Build GRCh38 | Loss |
|
| nsv1059098 | Chr.16:56733938 - 57028336 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map