Assay Details
Target Gene Details
Entrez Gene ID: | 220164 |
Gene Name: | docking protein 6 |
Gene Aliases: |
DOK5L, HsT3226 |
Location: |
Chr.18:69401027-69849087 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DOK6 | NM_152721.5 | NP_689934.2 | ||
| XM_011525875.2 | XP_011524177.1 | |||
| AY599248.1 | AAT09770.1 | |||
| BC096744.1 | AAH96744.1 | |||
| DB480182.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv32736 | Chr.18:69135149 - 71678611 on Build GRCh38 | Gain+Loss |
|
| nsv1072275 | Chr.18:69400763 - 69401364 on Build GRCh38 | Deletion |
|
| esv3642964 | Chr.18:69398892 - 69788959 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map