Assay Details
Target Gene Details
Entrez Gene ID: | 5912 |
Gene Name: | RAP2B, member of RAS oncogene family |
Gene Aliases: |
- |
Location: |
Chr.3:153162212-153170624 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RAP2B | NM_002886.3 | 1 | 5764 | NP_002877.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv998184 | Chr.3:152675973 - 153414946 on Build GRCh38 | Gain |
|
| nsv998158 | Chr.3:153167501 - 153222313 on Build GRCh38 | Gain |
|
| nsv954496 | Chr.3:153163812 - 153171511 on Build GRCh38 | Duplication |
|
| nsv820284 | Chr.3:153167653 - 153167957 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map