Assay Details
Target Gene Details
Entrez Gene ID: | 3981 |
Gene Name: | DNA ligase 4 |
Gene Aliases: |
LIG4S |
Location: |
Chr.13:108207442-108218368 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv1036n106 | Chr.13:108214752 - 108215552 on Build GRCh38 | Deletion |
|
| nsv1069770 | Chr.13:108214651 - 108215552 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map