Assay Details
Target Gene Details
Entrez Gene ID: | 80217 |
Gene Name: | cilia and flagella associated protein 43 |
Gene Aliases: |
C10orf79, WDR96, bA373N18.2 |
Location: |
Chr.10:104129888-104232363 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 25 - Exon 25 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CFAP43 | NM_025145.5 | NP_079421.5 | ||
| XM_005270171.2 | XP_005270228.1 | |||
| XM_005270172.3 | XP_005270229.1 | |||
| XM_011540196.2 | XP_011538498.1 | |||
| XM_011540197.2 | XP_011538499.1 | |||
| XM_011540198.2 | XP_011538500.1 | |||
| XM_011540199.2 | XP_011538501.1 | |||
| XM_011540200.2 | XP_011538502.1 | |||
| XM_011540201.2 | XP_011538503.1 | |||
| XM_011540202.2 | XP_011538504.1 | |||
| XM_011540203.2 | XP_011538505.1 | |||
| XM_017016681.1 | XP_016872170.1 | |||
| XM_017016682.1 | XP_016872171.1 | |||
| XM_017016683.1 | XP_016872172.1 | |||
| AL833836.1 | CAD38696.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831971 | Chr.10:104120100 - 104301309 on Build GRCh38 | Gain |
|

Genomic Map