Assay Details
Target Gene Details
Entrez Gene ID: | 200879 |
Gene Name: | lipase H |
Gene Aliases: |
AH, ARWH2, HYPT7, LAH2, LPDLR, PLA1B, mPA-PLA1 |
Location: |
Chr.3:185506262-185552661 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LIPH | NM_139248.2 | 10 | 2107 | NP_640341.1 |
| XM_006713529.3 | 9 | 2024 | XP_006713592.1 | |
| XM_011512530.2 | 11 | 2291 | XP_011510832.1 | |
| XM_011512531.2 | 12 | 2236 | XP_011510833.1 | |
| XM_017005852.1 | 9 | 2012 | XP_016861341.1 | |
| AK074229.1 | 9 | 1643 | BAB85023.1 | |
| AK122651.1 | ||||
| AY036912.1 | 10 | 2087 | AAK63178.1 | |
| AY093498.1 | 10 | 2079 | AAM18803.1 | |
| BC064941.1 | 10 | 2094 | AAH64941.1 | |
| BI965988.1 | 1 | 372 | ||
| EF186228.1 | 9 | 1982 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv829818 | Chr.3:185411745 - 185567640 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map