Assay Details
Target Gene Details
Entrez Gene ID: | 3178 |
Gene Name: | heterogeneous nuclear ribonucleoprotein A1 |
Gene Aliases: |
ALS19, ALS20, HNRPA1, HNRPA1L3, IBMPFD3, UP 1, hnRNP A1, hnRNP-A1 |
Location: |
Chr.12:54280690-54287088 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HNRNPA1 | NM_002136.3 | NP_002127.1 | ||
| NM_031157.3 | NP_112420.1 | |||
| NR_135167.1 | ||||
| XM_005268826.1 | XP_005268883.1 | |||
| XM_017019251.1 | XP_016874740.1 | |||
| AK129814.1 | ||||
| AK291113.1 | ||||
| AK298176.1 | ||||
| AK303303.1 | ||||
| AK308679.1 | ||||
| BC002355.2 | AAH02355.1 | |||
| BC009600.1 | AAH09600.1 | |||
| BC012158.1 | AAH12158.1 | |||
| BC033714.1 | AAH33714.1 | |||
| BC052296.1 | AAH52296.1 | |||
| BC070315.1 | AAH70315.1 | |||
| BC074502.1 | AAH74502.1 | |||
| BC103707.1 | AAI03708.1 | |||
| BC121133.2 | ||||
| BX425482.2 | ||||
| CD705336.1 | ||||
| DC358304.1 | ||||
| U00947.1 | ||||
| X06747.1 | CAA29922.1 | |||
| X79536.1 | CAA56072.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv25123 | Chr.12:54279681 - 54280827 on Build GRCh38 | Loss |
|
| nsv1049160 | Chr.12:54254119 - 54362754 on Build GRCh38 | Gain |
|
| nsv832420 | Chr.12:54137272 - 54296797 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map