Assay Details
Target Gene Details
Entrez Gene ID: | 93661 |
Gene Name: | capping actin protein of muscle Z-line alpha subunit 3 |
Gene Aliases: |
CAPPA3, Gsg3, HEL-S-86 |
Location: |
Chr.12:18738111-18739188 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CAPZA3 | NM_033328.2 | 1 | 969 | NP_201585.1 |
| AB053259.1 | 1 | 944 | BAB61901.1 | |
| AK058174.1 | 1 | 931 | BAB71703.1 | |
| BC016745.2 | 1 | 973 | AAH16745.1 | |
| GQ472212.1 | 1 | 973 |
More Information
Set Membership: |
|

Genomic Map