Assay Details
Target Gene Details
Entrez Gene ID: | 222643 |
Gene Name: | unc-5 family C-terminal like |
Gene Aliases: |
MUXA, ZUD |
Location: |
Chr.6:41026901-41039199 on Build GRCh38 |
Assay Gene Location: | Overlaps - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| UNC5CL | NM_173561.2 | NP_775832.2 | ||
| AI791395.1 | ||||
| BC035284.1 | AAH35284.1 | |||
| JF681947.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv526577 | Chr.6:41037763 - 41061603 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs74443078] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map