Assay Details
Target Gene Details
Entrez Gene ID: | 124925 |
Gene Name: | seizure related 6 homolog |
Gene Aliases: |
BSRPC |
Location: |
Chr.17:28954905-29006440 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 14 - Intron 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SEZ6 | NM_001098635.1 | NP_001092105.1 | ||
| NM_001290202.1 | NP_001277131.1 | |||
| NM_178860.4 | NP_849191.3 | |||
| XM_011524315.1 | XP_011522617.1 | |||
| XM_011524317.2 | XP_011522619.1 | |||
| AF502129.1 | AAM22213.1 | |||
| AF502130.1 | AAM22214.1 | |||
| AK054913.1 | BAB70826.1 | |||
| AK055383.1 | BAB70912.1 | |||
| AK091522.1 | BAC03684.1 | |||
| AK096473.1 | ||||
| AK125377.1 | ||||
| AK223620.1 | BAD97340.1 | |||
| AL834405.1 | CAD39067.1 | |||
| AY038048.1 | AAK71497.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv574662 | Chr.17:28867616 - 29014758 on Build GRCh38 | Loss |
|
| nsv833408 | Chr.17:28913470 - 29105450 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map