Assay Details
Target Gene Details
Entrez Gene ID: | 51092 |
Gene Name: | SID1 transmembrane family member 2 |
Gene Aliases: |
CGI-40 |
Location: |
Chr.11:117179223-117197445 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SIDT2 | NM_001040455.1 | 1 | 147 | NP_001035545.1 |
| AF151799.1 | 1 | 189 | AAD34036.1 | |
| AK292501.1 | 1 | 627 | ||
| AK292974.1 | 1 | 138 | ||
| AK303023.1 | 1 | 103 | ||
| AY358442.1 | 1 | 156 | AAQ88807.1 | |
| BC114522.1 | 1 | 176 | AAI14523.1 | |
| BC114959.1 | AAI14960.1 | |||
| KC109742.1 | 1 | 103 |
More Information
Set Membership: |
|

Genomic Map