Assay Details
Target Gene Details
Entrez Gene ID: | 84950 |
Gene Name: | pre-mRNA processing factor 38A |
Gene Aliases: |
Prp38 |
Location: |
Chr.1:52404547-52419723 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PRPF38A | NM_032864.3 | 1 | 72 | NP_116253.2 |
| XM_011542315.2 | 1 | 72 | XP_011540617.1 | |
| AK092038.1 | 1 | 55 | BAC03796.1 | |
| AK315621.1 | 1 | 56 | ||
| BC000777.2 | AAH00777.2 | |||
| BC063655.1 | 1 | 48 | AAH63655.1 | |
| DA055001.1 | 1 | 72 |
More Information
Set Membership: |
|

Genomic Map