Assay Details
Target Gene Details
Entrez Gene ID: | 10633 |
Gene Name: | RAS like family 10 member A |
Gene Aliases: |
RRP22 |
Location: |
Chr.22:29312933-29319618 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RASL10A | NM_006477.4 | 3 | 1218 | NP_006468.1 |
| XM_011529821.1 | 4 | 936 | XP_011528123.1 | |
| XM_011529822.1 | 4 | 1692 | XP_011528124.1 | |
| XM_011529823.1 | 3 | 799 | XP_011528125.1 | |
| BC022473.1 | 3 | 1218 | AAH22473.1 | |
| BC040434.1 | 2 | 1242 | AAH40434.1 | |
| BC050739.1 | 2 | 1448 | AAH50739.1 | |
| BC058077.1 | 3 | 1127 | AAH58077.1 | |
| BX111994.1 | 1 | 265 | ||
| Y07847.1 | 3 | 1029 | CAA69175.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834171 | Chr.22:29230413 - 29433855 on Build GRCh38 | Loss |
|
| nsv829154 | Chr.22:29221496 - 29328012 on Build GRCh38 | Gain |
|
| nsv829155 | Chr.22:29268407 - 29328012 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map