Assay Details
Target Gene Details
Entrez Gene ID: | 57001 |
Gene Name: | succinate dehydrogenase complex assembly factor 3 |
Gene Aliases: |
ACN9, DC11, LYRM10, Sdh7 |
Location: |
Chr.7:97116593-97181763 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SDHAF3 | NM_020186.2 | 2 | 1356 | NP_064571.1 |
| AF201933.1 | 2 | 245 | AAF86869.1 | |
| BC020621.1 | 2 | 238 | AAH20621.1 | |
| BC022858.1 | 2 | 246 | AAH22858.1 |
More Information
Set Membership: |
|

Genomic Map