Assay Details
Target Gene Details
Entrez Gene ID: | 553 |
Gene Name: | arginine vasopressin receptor 1B |
Gene Aliases: |
AVPR3 |
Location: |
Chr.1:206109849-206117048 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AVPR1B | NM_000707.3 | NP_000698.1 | ||
| D31833.1 | BAA06621.1 | |||
| L37112.1 | 2 | 1713 | AAA65687.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832414 | Chr.1:206069379 - 206172035 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map