Assay Details
Target Gene Details
Entrez Gene ID: | 433 |
Gene Name: | asialoglycoprotein receptor 2 |
Gene Aliases: |
ASGP-R2, ASGPR2, CLEC4H2, HBXBP, HL-2 |
Location: |
Chr.17:7101322-7115522 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ASGR2 | NM_001181.4 | NP_001172.1 | ||
| NM_001201352.1 | NP_001188281.1 | |||
| NM_080912.3 | NP_550434.1 | |||
| NM_080913.3 | NP_550435.1 | |||
| NM_080914.2 | NP_550436.1 | |||
| XM_005256648.2 | XP_005256705.1 | |||
| XM_006721524.2 | XP_006721587.2 | |||
| XM_006721526.1 | XP_006721589.1 | |||
| XM_011523862.2 | XP_011522164.2 | |||
| XM_011523863.2 | XP_011522165.1 | |||
| XM_011523864.2 | XP_011522166.1 | |||
| XM_011523865.1 | XP_011522167.1 | |||
| XM_011523866.1 | XP_011522168.1 | |||
| XM_017024651.1 | XP_016880140.1 | |||
| XM_017024652.1 | XP_016880141.1 | |||
| XM_017024653.1 | XP_016880142.1 | |||
| AF529374.1 | AAQ09608.1 | |||
| BC017251.2 | AAH17251.1 | |||
| M11025.1 | AAB59519.1 | |||
| U97197.1 | AAB58308.1 | |||
| X55283.1 | CAA38997.1 | |||
| X55284.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2422288 | Chr.17:6789933 - 7646709 on Build GRCh38 | Deletion |
|
| esv2665200 | Chr.17:7091816 - 7333062 on Build GRCh38 | Deletion |
|
| nsv574322 | Chr.17:7010333 - 7689462 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map