Assay Details
Target Gene Details
Entrez Gene ID: | 100526760 |
Gene Name: | ABHD14A-ACY1 readthrough |
Gene Aliases: |
- |
Location: |
Chr.3:51975026-51989202 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ABHD14A-ACY1 | NM_001316331.1 | 4 | 565 | NP_001303260.1 |
| AK294015.1 | 2 | 203 | ||
| AK298086.1 | 4 | 541 |
Target Gene Details
Entrez Gene ID: | 95 |
Gene Name: | aminoacylase 1 |
Gene Aliases: |
ACY-1, ACY1D, HEL-S-5 |
Location: |
Chr.3:51983284-51989202 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ACY1 | NM_000666.2 | 2 | 364 | NP_000657.1 |
| NM_001198895.1 | 2 | 359 | NP_001185824.1 | |
| NM_001198896.1 | 2 | 364 | NP_001185825.1 | |
| NM_001198897.1 | 2 | 364 | NP_001185826.1 | |
| NM_001198898.1 | 2 | 364 | NP_001185827.1 | |
| AK298274.1 | 2 | 113 | ||
| BC000545.2 | 2 | 108 | AAH00545.1 | |
| BC003023.1 | 2 | 101 | AAH03023.1 | |
| BC014112.2 | 2 | 84 | AAH14112.1 | |
| BE269642.1 | 2 | 364 | ||
| CD014023.1 | 2 | 205 | ||
| CD014026.1 | 2 | 205 | ||
| CD014027.1 | 2 | 205 | ||
| D14524.1 | 2 | 98 | BAA03397.1 | |
| D16307.1 | 2 | 96 | BAA03814.1 | |
| EU794586.1 | 2 | 84 | ||
| KU177890.1 | ||||
| KU177891.1 | ||||
| L07548.1 | 2 | 101 | AAA02852.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1003453 | Chr.3:51928065 - 52065425 on Build GRCh38 | Gain |
|
| nsv508922 | Chr.3:51940932 - 52009520 on Build GRCh38 | Insertion |
|
| nsv954857 | Chr.3:51932485 - 52056584 on Build GRCh38 | Deletion |
|
| nsv519445 | Chr.3:51970301 - 52333916 on Build GRCh38 | Gain+Loss |
|
| nsv520211 | Chr.3:51738331 - 52132131 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map