Assay Details
Target Gene Details
Entrez Gene ID: | 3882 |
Gene Name: | keratin 32 |
Gene Aliases: |
HA2, HKA2, KRTHA2, hHa2 |
Location: |
Chr.17:41459513-41469252 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KRT32 | NM_002278.3 | 7 | 1413 | NP_002269.3 |
| X81419.1 | 7 | 1252 | CAA57179.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv510711 | Chr.17:41455775 - 41574964 on Build GRCh38 | Deletion |
|
| nsv483014 | Chr.17:41339595 - 41514244 on Build GRCh38 | Loss |
|
| esv2758690 | Chr.17:41145123 - 41740591 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map