Assay Details
Target Gene Details
Entrez Gene ID: | 10500 |
Gene Name: | semaphorin 6C |
Gene Aliases: |
SEMAY, m-SemaY, m-SemaY2 |
Location: |
Chr.1:151131685-151148408 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 6 - Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SEMA6C | NM_001178061.1 | NP_001171532.1 | ||
| NM_001178062.1 | NP_001171533.1 | |||
| NM_030913.4 | NP_112175.2 | |||
| XM_005244835.3 | XP_005244892.1 | |||
| XM_017000075.1 | XP_016855564.1 | |||
| XM_017000076.1 | XP_016855565.1 | |||
| XM_017000077.1 | XP_016855566.1 | |||
| XM_017000078.1 | XP_016855567.1 | |||
| XM_017000079.1 | XP_016855568.1 | |||
| XM_017000080.1 | XP_016855569.1 | |||
| XM_017000081.1 | XP_016855570.1 | |||
| AB022434.1 | BAB20670.1 | |||
| AB058772.1 | ||||
| AF339152.1 | AAL72098.1 | |||
| AF339153.1 | AAL72099.1 | |||
| AF339154.1 | AAL72100.1 | |||
| AK302862.1 | ||||
| AK311613.1 | ||||
| BC114521.1 | AAI14522.1 | |||
| BC114623.1 | AAI14624.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv522840 | Chr.1:150961247 - 151375270 on Build GRCh38 | Gain |
|
| dgv2n68 | Chr.1:151136983 - 151309302 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map