Assay Details
Target Gene Details
Entrez Gene ID: | 283579 |
Gene Name: | chromosome 14 open reading frame 178 |
Gene Aliases: |
- |
Location: |
Chr.14:77760830-77769742 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C14orf178 | NM_001173978.1 | 1 | 95 | NP_001167449.1 |
| NM_174943.3 | 1 | 95 | NP_777603.1 | |
| AK098842.1 | 2 | 340 | BAC05432.1 |
Target Gene Details
Entrez Gene ID: | 22938 |
Gene Name: | SNW domain containing 1 |
Gene Aliases: |
Bx42, NCOA-62, PRPF45, Prp45, SKIIP, SKIP |
Location: |
Chr.14:77717599-77761220 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SNW1 | NM_001318844.1 | NP_001305773.1 | ||
| NM_012245.2 | NP_036377.1 | |||
| XM_005267413.3 | XP_005267470.1 | |||
| AF045184.1 | AAC31697.1 | |||
| AK292274.1 | ||||
| AK293619.1 | ||||
| AK302246.1 | ||||
| AK316087.1 | ||||
| AX775981.1 | CAE11690.1 | |||
| BC105585.1 | ||||
| BC108903.1 | AAI08904.1 | |||
| U51432.1 | AAC15912.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759998 | Chr.14:77607837 - 77796229 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map