Assay Details
Target Gene Details
Entrez Gene ID: | 79833 |
Gene Name: | gem nuclear organelle associated protein 6 |
Gene Aliases: |
- |
Location: |
Chr.2:38778185-38781965 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GEMIN6 | NM_024775.9 | NP_079051.9 | ||
| AF453443.1 | AAL48292.1 | |||
| AK027112.1 | BAB15660.1 | |||
| AK315627.1 | ||||
| BC018195.1 | AAH18195.1 | |||
| BM471707.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv9646 | Chr.2:38131169 - 38855047 on Build GRCh38 | Loss |
|
| nsv1014616 | Chr.2:38758330 - 38795751 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map