Assay Details
Target Gene Details
Entrez Gene ID: | 2867 |
Gene Name: | free fatty acid receptor 2 |
Gene Aliases: |
FFA2R, GPR43 |
Location: |
Chr.19:35447965-35451767 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FFAR2 | NM_005306.2 | NP_005297.1 | ||
| XM_017026709.1 | XP_016882198.1 | |||
| XM_017026710.1 | XP_016882199.1 | |||
| XM_017026711.1 | XP_016882200.1 | |||
| AB378083.1 | ||||
| AX810701.1 | ||||
| BC095535.1 | AAH95535.1 | |||
| BC096198.3 | AAH96198.1 | |||
| BC096199.3 | AAH96199.1 | |||
| BC096200.1 | AAH96200.1 | |||
| BC096201.1 | AAH96201.1 | |||
| KP844446.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833811 | Chr.19:35359355 - 35538633 on Build GRCh38 | Gain |
|
| nsv833810 | Chr.19:35291685 - 35481057 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map