Assay Details
Target Gene Details
Entrez Gene ID: | 6804 |
Gene Name: | syntaxin 1A |
Gene Aliases: |
HPC-1, P35-1, STX1, SYN1A |
Location: |
Chr.7:73699205-73719702 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| STX1A | NM_001165903.1 | NP_001159375.1 | ||
| NM_004603.3 | NP_004594.1 | |||
| XM_017012567.1 | XP_016868056.1 | |||
| AB086954.1 | BAC78519.1 | |||
| AK299959.1 | ||||
| AK313980.1 | ||||
| BC003011.1 | AAH03011.1 | |||
| BC064644.1 | AAH64644.1 | |||
| CR457018.1 | CAG33299.1 | |||
| D37932.1 | BAA07151.1 | |||
| DA159446.1 | ||||
| KJ534952.1 | ||||
| KJ534980.1 | ||||
| L37792.1 | AAA53519.1 | |||
| U12918.1 | AAA20940.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831028 | Chr.7:73556313 - 73816595 on Build GRCh38 | Loss |
|
| nsv469841 | Chr.7:73621181 - 73770495 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map