Assay Details
Target Gene Details
Entrez Gene ID: | 6500 |
Gene Name: | S-phase kinase-associated protein 1 |
Gene Aliases: |
EMC19, OCP-II, OCP2, SKP1A, TCEB1L, p19A |
Location: |
Chr.5:134156391-134177033 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SKP1 | NM_006930.3 | NP_008861.2 | ||
| NM_170679.2 | NP_733779.1 | |||
| AK057110.1 | 1 | 752 | ||
| BC009839.2 | AAH09839.1 | |||
| BC020798.1 | AAH20798.1 | |||
| BC025673.1 | AAH25673.1 | |||
| BC065730.1 | AAH65730.1 | |||
| BI552966.1 | ||||
| BI670085.1 | ||||
| U33760.1 | AAC50241.1 | |||
| U37558.1 | AAA79202.1 | |||
| Z47087.1 | CAA87392.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv599732 | Chr.5:134099223 - 134230401 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map