Assay Details
Target Gene Details
Entrez Gene ID: | 10512 |
Gene Name: | semaphorin 3C |
Gene Aliases: |
SEMAE, SemE |
Location: |
Chr.7:80742538-80919351 on Build GRCh38 |
Assay Gene Location: | Within Exon 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SEMA3C | NM_006379.3 | 18 | 4695 | NP_006370.1 |
| XM_005250113.2 | 19 | 4616 | XP_005250170.1 | |
| AB000220.1 | 18 | 4695 | BAA32398.1 | |
| BC030690.1 | 18 | 4498 | AAH30690.1 |
More Information
Set Membership: |
|

Genomic Map