Assay Details
Target Gene Details
Entrez Gene ID: | 57506 |
Gene Name: | mitochondrial antiviral signaling protein |
Gene Aliases: |
CARDIF, IPS-1, IPS1, VISA |
Location: |
Chr.20:3846799-3876123 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MAVS | NM_001206491.1 | NP_001193420.1 | ||
| NM_020746.4 | NP_065797.2 | |||
| NR_037921.1 | ||||
| AB097003.1 | BAC77356.1 | |||
| AK023799.1 | BAB14684.1 | |||
| AK123956.1 | BAC85734.1 | |||
| AK130992.1 | BAC85473.1 | |||
| AK291785.1 | ||||
| AK296897.1 | ||||
| BC044952.1 | AAH44952.1 | |||
| BM834250.1 | ||||
| BX649084.1 | ||||
| DB219666.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833899 | Chr.20:3709295 - 3878350 on Build GRCh38 | Loss |
|
| nsv522507 | Chr.20:3846662 - 3849827 on Build GRCh38 | Loss |
|
| nsv585302 | Chr.20:3842371 - 3849827 on Build GRCh38 | Gain |
|
| nsv828673 | Chr.20:3738382 - 4184811 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map