Assay Details
Target Gene Details
Entrez Gene ID: | 8153 |
Gene Name: | Rho family GTPase 2 |
Gene Aliases: |
ARHN, RHO7, RhoN |
Location: |
Chr.17:43025241-43032041 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 5 - Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RND2 | NM_005440.4 | NP_005431.1 | ||
| XM_011525316.1 | XP_011523618.1 | |||
| XM_011525317.2 | XP_011523619.1 | |||
| AB209778.1 | BAD93015.1 | |||
| AF498968.1 | AAM21115.1 | |||
| AK290199.1 | ||||
| BC094842.1 | AAH94842.1 | |||
| BC104986.1 | AAI04987.1 | |||
| BC104990.1 | AAI04991.1 | |||
| BT019394.1 | AAV38201.1 | |||
| DB502558.1 | ||||
| X95456.1 | CAA64726.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2672913 | Chr.17:42964983 - 43322347 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map