Assay Details
Target Gene Details
Entrez Gene ID: | 3313 |
Gene Name: | heat shock protein family A (Hsp70) member 9 |
Gene Aliases: |
CRP40, CSA, EVPLS, GRP-75, GRP75, HEL-S-124m, HSPA9B, MOT, MOT2, MTHSP75, PBP74, SAAN, SIDBA4 |
Location: |
Chr.5:138554882-138575629 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 10 - Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HSPA9 | NM_004134.6 | NP_004125.3 | ||
| AK023317.1 | 1 | 973 | ||
| AK222758.1 | BAD96478.1 | |||
| AK225488.1 | ||||
| AK293990.1 | ||||
| AK297795.1 | ||||
| AK297897.1 | ||||
| AK315177.1 | ||||
| BC000478.2 | AAH00478.1 | |||
| BC024034.2 | AAH24034.1 | |||
| BC030634.1 | AAH30634.1 | |||
| DQ480334.1 | ||||
| FJ224291.1 | ||||
| L11066.1 | ||||
| L15189.1 | AAA67526.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv599818 | Chr.5:138559529 - 138641163 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map